missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure
Use directly to resuspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
$130.41 - $188.81
Chemical Identifiers
| CAS | 1185-53-1 |
|---|---|
| Molecular Formula | C4H12ClNO3 |
| Molecular Weight (g/mol) | 157.594 |
| InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
| PubChem CID | 93573 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| SMILES | C(C(CO)(CO)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ75893XCR
|
Thermo Fisher Scientific
J75893XCR |
10 x 1 mL | Nalgene bottle |
Case of 10 Each for $130.41
|
|
||||
|
AAJ75893AE
|
Thermo Fisher Scientific
J75893AE |
100 mL | Nalgene bottle |
Each for $152.97
|
|
||||
|
AAJ75893AP
|
Thermo Fisher Scientific
J75893AP |
500 mL | Nalgene bottle |
Each for $188.81
|
|
||||
Description
Use directly to resuspend and/or dilute purified DNA or RNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1185-53-1 | |
| 157.594 | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
| C4H12ClNO3 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
| 1185-53-1 | |
| 10 x 1 mL | |
| Nalgene bottle | |
| C(C(CO)(CO)N)O.Cl | |
| 157.594 | |
| Molecular Biology |
| Solution | |
| C4H12ClNO3 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 | |
| TE Buffer |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only