missing translation for 'onlineSavingsMsg'
Learn More

TE Buffer, 1X Solution, Molecular Biology Grade, Ultrapure

Use directly to resuspend and/or dilute purified DNA or RNA | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Supplier:  Thermo Fisher Scientific J75893AE

 View more versions of this product

Catalog No. AAJ75893AE


Only null left

Description

Description

Use directly to resuspend and/or dilute purified DNA or RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

TE Buffer
Solution
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
1185-53-1
100 mL
Nalgene bottle
C(C(CO)(CO)N)O.Cl
157.594
Molecular Biology

Chemical Identifiers

1185-53-1
157.594
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only