missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure
Used in Western blotting protocols | CAS: 7647-14-5 | ClNa | 58.44 g/mol
$128.88 - $437.85
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium;chloride |
| SMILES | [Na+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAJ75892AE
|
Thermo Fisher Scientific
J75892AE |
100 mL | Nalgene bottle |
Each for $128.88
|
|
||||
|
AAJ75892K2
|
Thermo Fisher Scientific
J75892K2 |
1 L | Nalgene bottle |
Each for $246.40
|
|
||||
|
AAJ75892K8
|
Thermo Fisher Scientific
J75892K8 |
5 L | Upright carton |
Each for $437.85
|
|
||||
Description
Used in Western blotting protocols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| 5234 | |
| sodium;chloride |
Specifications
| 7647-14-5 | |
| Western blotting protocols | |
| Solution | |
| ClNa | |
| Nalgene bottle | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| TBS |
| 20X | |
| 7.4 | |
| 100 mL | |
| MFCD00003477 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium;chloride | |
| 5234 | |
| Molecular Biology, Ultrapure |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only