missing translation for 'onlineSavingsMsg'
Learn More

TBS, 20X Solution, pH 7.4, Molecular Biology Grade, Ultrapure

Used in Western blotting protocols | CAS: 7647-14-5 | ClNa | 58.44 g/mol

Supplier:  Thermo Fisher Scientific J75892K2

 View more versions of this product

Catalog No. AAJ75892K2


Only null left
Explore available promotions

Description

Description

Used in Western blotting protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

TBS
20X
7.4
1 L
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Molecular Biology, Ultrapure
7647-14-5
Western blotting protocols
Solution
ClNa
Nalgene bottle
[Na+].[Cl-]
58.44
CHEBI:26710

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only