missing translation for 'onlineSavingsMsg'
Learn More

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™

Catalog No. BP13334
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP13334 4 L Poly CUBE
BP13331 1 L Nalgene Poly Bottle
BP133320 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP13334 Supplier Fisher BioReagents Supplier No. BP13334
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-35-3

TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TBE buffer solution - simply dilute as needed
  • 10X solution contains 0.89M tris, 0.89M borate, and 0.02M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBE Solution with 9 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Functionally tested for use in electrophoresis applications
  • TBE is a commonly used buffer for electrophoresis of nucleic acids in agarose and polyacrylamide gels
  • Intended for laboratory research use only

Chemical Identifiers

CAS 10043-35-3 , 60-00-4 , 77-86-1 , 7732-18-5
Synonym TBE

Specifications

Color Colorless
pH 8.2 to 8.4
Physical Form Liquid
Quantity 4 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Packaging Poly CUBE
Protease Protease free
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris-Borate-EDTA, 10X Solution
Safety handling Check Alert storage code Red
Recommended Storage RT