missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 3'Primer Modification | None |
|---|---|
| 5'Primer Modification | None |
| Content And Storage | 1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers). Store at -15 to -25°C. |
| Description | VIC™ - MGB probe detects Allele 1, FAM - MGB probe detects Allele 2 |
| Form | Frozen |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
43-513-84
|
Thermo Fisher Scientific
4351384 |
S (300 reactions), Made to Order |
Each for $690.00
|
|
|||||
|
43-513-82
|
Thermo Fisher Scientific
4351382 |
M (1000 reactions), Made to Order |
Each for $1,254.00
|
|
|||||
|
43-513-80
|
Thermo Fisher Scientific
4351380 |
L (2400 reactions), Made to Order |
Each for $2,185.00
|
|
|||||
Specifications
| None | |
| 1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers). Store at -15 to -25°C. |
|
| Frozen | |
| 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler* * If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals. |
|
| FAM, VIC | |
| Solid Phase Extraction (SPE) | |
| Room Temperature | |
| Over 10,000 mouse assays are available |
| None | |
| VIC™ - MGB probe detects Allele 1, FAM - MGB probe detects Allele 2 | |
| Made to Order | |
| VIC™ (5'), FAM (5'), MGB (Minor Groove Binder) (3') | |
| TaqMan | |
| RNase-Free, DNase-Free | |
| Mouse | |
| Genotyping |