missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TaqMan™ SNP Genotyping Assay, mouse
Specifications
Specifications
| 3'Primer Modification | None |
| Concentration | 40X |
| 5'Primer Modification | None |
| Content And Storage | 1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers). Store at -15 to -25°C. |
| Description | VIC™ - MGB probe detects Allele 1, FAM - MGB probe detects Allele 2 |
| Form | Frozen |
| Format | Made to Order |
| For Use With (Equipment) | 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler* * If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals. |
| Internal Probe Modification | VIC™ (5'), FAM (5'), MGB (Minor Groove Binder) (3') |
| Label or Dye | FAM, VIC |
| Show More |