Learn More
TaqMan™ SNP Genotyping Assay, mouse
Supplier:  Thermo Fisher Scientific 4351380
 View more versions of this product
Catalog No. 43-513-80
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
None
None
VIC™ - MGB probe detects Allele 1, FAM - MGB probe detects Allele 2
Made to Order
Sustainable packaging
FAM, VIC
Solid Phase Extraction (SPE)
L (2400 reactions), Made to Order
Mouse
Genotyping
80X
1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).
Store at -15 to -25°C.
Frozen
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler*
* If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR
system in order to detect and record fluorescent signals.
VIC™ (5'), FAM (5'), MGB (Minor Groove Binder) (3')
TaqMan
RNase-Free, DNase-Free
Room Temperature
Over 10,000 mouse assays are available
2400 reactions
Product Suggestions
Product Suggestions