missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 0.2M buffer soln., pH 8.0

CAS: 91000-53-2265.256 g/mol

$165.23 - $254.18

Chemical Identifiers

CAS 91000-53-2
Molecular Weight (g/mol) 265.256
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60322AE
View Documents
Thermo Scientific Chemicals
J60322AE
100 mL
Each for $165.23
Only null left
 
AAJ60322AK
View Documents
Thermo Scientific Chemicals
J60322AK
250 mL
Each for $254.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

91000-53-2
265.256

Specifications

91000-53-2
8.0
100 mL
TAPS
0.2 M
Liquid
265.256
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only