CAS RN 91000-53-2
IUPAC Name:
sodium 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonate
Synonyms:
sodium 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonate
Molecular Weight (g/mol):
265.26
Molecular Formula:
C7H16NNaO6S
InChi Key:
FEGYIWVHCSRXCG-UHFFFAOYSA-M
SMILES:
[Na+].OCC(CO)(CO)NCCCS([O-])(=O)=O
1
–
3
of
3
results
1
Thermo Scientific Chemicals TAPS, 0.2M buffer soln., pH 8.0
CAS: 91000-53-2 Molecular Weight (g/mol): 265.256
2
TAPS sodium salt, 98%
CAS: 91000-53-2 Molecular Formula: C7H16NNaO6S Molecular Weight (g/mol): 265.256 MDL Number: MFCD00069903 InChI Key: FEGYIWVHCSRXCG-UHFFFAOYSA-M PubChem CID: 23667519 IUPAC Name: sodium;3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonate SMILES: C(CNC(CO)(CO)CO)CS(=O)(=O)[O-].[Na+]
3
TAPS, 0.2M buffer soln., pH 8.5, Thermo Scientific Chemicals
CAS: 91000-53-2 Molecular Weight (g/mol): 265.256