missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 0.2M buffer soln., pH 8.0

CAS: 91000-53-2265.256 g/mol

Supplier:  Thermo Scientific Chemicals J60322AK

 View more versions of this product

Catalog No. AAJ60322AK


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

TAPS
0.2 M
Liquid
265.256
91000-53-2
8.0
250 mL

Chemical Identifiers

91000-53-2
265.256
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only