Learn More
Synthware™ West Condenser, with Removable Hose Connections
GREENER_CHOICE
Has narrow annular space between the inner tube and jacket that provides high cooling efficiency
$175.46 - $252.00
Specifications
Material Glass
Type West Condenser
View More Specs
Products 6
Catalog Number Mfr. No. Standard Taper Joint No Length (Metric) Jacket Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint No Length (Metric) Jacket Price Quantity & Availability  
31500769
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C551110
14/20 110 mm
Each for $175.46
Only null left
31500770
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C550190
14/20 190 mm
Each for $188.82
Only null left
31500771
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C554100
24/40 100 mm
Each for $214.18
Only null left
31500773
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C554200
24/40 200 mm
Each for $238.00
Only null left
31500772
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C551190
24/40 190 mm
Each for $230.25
Only null left
31500774
GREENER_CHOICE
View Documents Promotion Details
Synthware™
C554300
24/40 300 mm
Each for $252.00
Only null left
Description
Description
  • Top standard taper outer joint and lower drip tip joint of same size
Specifications
Specifications
Glass
West Condenser