Learn More
Synthware™ West Condenser, with Removable Hose Connections
GREENER_CHOICE
Has narrow annular space between the inner tube and jacket that provides high cooling efficiency
Supplier:  Synthware™ C550190
 View more versions of this product
Catalog No. 31500770
Only null left
Description
Description
  • Top standard taper outer joint and lower drip tip joint of same size
Specifications
Chemical Identifiers
Glass
West Condenser
Specifications
Glass
14/20
190 mm
West Condenser
Product Suggestions
Product Suggestions