Learn More
Synthware™ West Condenser, with Removable Hose Connections
Description
- Top standard taper outer joint and lower drip tip joint of same size
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Length (Metric) Jacket | 300 mm |
| Standard Taper Joint No | 24/40 |
| Type | West Condenser |