Learn More
Synthware™ West Condenser, with Removable Hose Connections
Has narrow annular space between the inner tube and jacket that provides high cooling efficiency
Supplier: Synthware™ C554200
Description
- Top standard taper outer joint and lower drip tip joint of same size
Chemical Identifiers
| Glass | |
| West Condenser |
Specifications
| Glass | |
| 200 mm | |
| 24/40 | |
| West Condenser |