Learn More
Synthware™ West Condenser, with Removable Hose Connections
GREENER_CHOICE
Has narrow annular space between the inner tube and jacket that provides high cooling efficiency
Supplier:  Synthware™ C554100
 View more versions of this product
Catalog No. 31500771
Only null left
Description
Description
  • Top standard taper outer joint and lower drip tip joint of same size
Specifications
Chemical Identifiers
Glass
West Condenser
Specifications
Glass
West Condenser
100 mm
24/40
Product Suggestions
Product Suggestions