Learn More
Synthware™ Distillation Receiver, Dean Stark
For easy separation of water from reaction or distillation system
$413.50 - $474.50
| Catalog Number | Mfr. No. | Standard Taper Joint | Capacity | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint | Capacity | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||
31501062
![]() |
Synthware™
D451414 |
14/20 | 5 mL | 205mm | 65mm |
Each for $413.50
|
|
||
31501064
![]() |
Synthware™
D451910 |
19/22 | 10 mL | 190mm | 75mm |
Each for $446.68
|
|
||
31501063
![]() |
Synthware™
D451410 |
14/20 | 10 mL | 190mm | 65mm |
Each for $436.46
|
|
||
31501065
![]() |
Synthware™
D452410 |
24/40 | 10 mL | 275mm | 95mm |
Each for $457.04
|
|
||
31501067
![]() |
Synthware™
D452910 |
29/42 | 10 mL | 275mm | 95mm |
Each for $458.96
|
|
||
31501068
![]() |
Synthware™
D452920 |
29/42 | 20 mL | 325mm | 95mm |
Each for $474.50
|
|
||
31501066
![]() |
Synthware™
D452424 |
24/40 | 20 mL | 325mm | 95mm |
Each for $473.32
|
|
||
Description
- Graduated
- With upper outer and lower inner standard taper joints, both of same size
- PTFE stopcock with 2 mm bore
Specifications
| Distillation Receiver |