Learn More
Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Polyurethane Coating
Cut-resistant gloves feature composite 13-gauge knit and polyurethane palm coating for secure grip and tactility.
$13.36 - $13.36
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE
Coating Material Polyurethane
Color Cream, Gray
Cut Resistance Rating ANSI A4
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-400-186
View Documents
Superior Glove™
S13KFGPU8
8
Pair for $13.36
Case of 240 Pair for $2,844.00
Only null left
17-400-187
View Documents
Superior Glove™
S13KFGPU9
9
Pair for $13.36
Case of 240 Pair for $2,844.00
Only null left
17-400-188
View Documents
Superior Glove™
S13KFGPU10
10
Pair for $13.36
Case of 240 Pair for $1,228.80
Only null left
17-400-189
View Documents
Superior Glove™
S13KFGPU11
11
Pair for $13.36
Case of 240 Pair for $2,844.00
Only null left
Description
Description
  • Composite fine-gauge knit is highly dexterous
  • Cut-resistant, abrasion resistant, puncture resistant
  • Polyurethane palm coating provides secure grip and tactility
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Dipped over an ergonomic hand form for a natural, comfortable fit
Specifications
Specifications
ANSI 4
Polyurethane
ANSI A4
Cut-resistant Gloves
automotive, light Metal stamping, fabrication, small parts handling assembly, Glass handling, sheet Metal work
ANSI/ISEA 105-16 Level 2
13
CE
Cream, Gray
Aramid Fiber, Silica
EN 388:4X31B
Knit
Dexterity™
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'