missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Polyurethane Coating

Catalog No. 17400186
Change view
Click to view available options
Size:
10
11
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17400186 8
17400187 9
17400188 10
17400189 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17400186 Supplier Superior Glove™ Supplier No. S13KFGPU/8

Cut-resistant gloves feature composite 13-gauge knit and polyurethane palm coating for secure grip and tactility.

  • Composite fine-gauge knit is highly dexterous
  • Cut-resistant, abrasion resistant, puncture resistant
  • Polyurethane palm coating provides secure grip and tactility
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Dipped over an ergonomic hand form for a natural, comfortable fit

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE
Coating Material Polyurethane
Color Cream, Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Silica
Product Type Cut-resistant Gloves
Size 8
EN388 Standard EN 388:4X31B
For Use With (Application) automotive, light Metal stamping, fabrication, small parts handling assembly, Glass handling, sheet Metal work
Glove Cuff Style Knit
Model S13KFGPU/8
Puncture Rating ANSI/ISEA 105-16 Level 2
Series Dexterity™
Yarn Gauge 13
Show More Show Less