Learn More
Superior Glove™ Dexterity™ Cut-Resistant String Knit Gloves, Polyurethane Coating
Cut-resistant gloves feature composite 13-gauge knit and polyurethane palm coating for secure grip and tactility.
Supplier:  Superior Glove™ S13KFGPU11
 View more versions of this product
Catalog No. 17-400-189
Only null left
Explore available promotions
Description
Description
  • Composite fine-gauge knit is highly dexterous
  • Cut-resistant, abrasion resistant, puncture resistant
  • Polyurethane palm coating provides secure grip and tactility
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Dipped over an ergonomic hand form for a natural, comfortable fit
Specifications
Specifications
ANSI 4
Polyurethane
ANSI A4
Cut-resistant Gloves
EN 388:4X31B
Knit
ANSI/ISEA 105-16 Level 2
13
CE
Cream, Gray
Aramid Fiber, Silica
11
automotive, light Metal stamping, fabrication, small parts handling assembly, Glass handling, sheet Metal work
S13KFGPU/11
Dexterity™
Product Suggestions
Product Suggestions