missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 10% (w/v), Ricca Chemical

$139.20 - $139.20

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
8125-16
View Documents
Ricca Chemical Company
812516
500 mL Amber Glass Bottle
Each for $139.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5965-83-3
250.27
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2723734
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
C9H14O6S
MFCD00007508,MFCD00149540
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
2-hydroxy-5-sulfobenzoic acid; bis(methane)

Specifications

5965-83-3 , 7732-18-5
93.0,9.28
MFCD00007508,MFCD00149540
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid; bis(methane)
2723734
Laboratory
Amber Glass Bottle
500 mL
89.0,8.91
C9H14O6S
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
250.27
10%
10%(w/v)
Aqueous solution
Sulfosalicylic Acid
Videos
SDS
Documents

Documents