missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine USP , Medisca™

$125.73 - $1763.43

Chemical Identifiers

CAS 599-79-1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-028
View Documents
Medisca Pharmaceutique Inc
017604
25 g
Each for $125.73
Only null left
 
18-048-029
View Documents
Medisca Pharmaceutique Inc
017609
1 kg
Each for $1,763.43
Only null left
 
18-048-084
View Documents
Medisca Pharmaceutique Inc
017605
100 g
Each for $352.69
Only null left
 
18-048-124
View Documents
Medisca Pharmaceutique Inc
017608
500 g
Each for $1,073.40
Only null left
 
Description

Description

Sulfasalazine USP is a bright yellow or brownish yellow, odorless, fine powder.

Specifications

Chemical Identifiers

599-79-1

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
599-79-1
25 g
USP
Powder
Brown to Yellow
97.0 to 101.5%
Videos
SDS
Documents

Documents