Learn More
Sulfasalazine USP , Medisca™
Supplier: Medisca Pharmaceutique Inc 017609
Description
Sulfasalazine USP is a bright yellow or brownish yellow, odorless, fine powder.
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 599-79-1 | |
| 1 kg | |
| USP |
| Powder | |
| Brown to Yellow | |
| 97.0 to 101.5% |
Chemical Identifiers
| 599-79-1 |