missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide Reagent (1% w/v) in 10% v/v Hydrochloric Acid, Reagents

Catalog No. 01333960
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333960 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333960 Supplier Labchem Inc Supplier No. CS0118001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfanilamide Reagent (1% w/v) in 10% v/v Hydrochloric Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7647-01-0 , 63-74-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulfamine, 4-Aminobenzenesulfonamide
IUPAC Name water
SMILES O

Specifications

Color Colorless
pH 0
Boiling Point 103°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1.0197
Grade Reagent
Physical Form Liquid
Chemical Name or Material Sulfanilamide Reagent (1% w/v) in 10% v/v Hydrochloric Acid