Learn More
Sulfanilamide, For Nitrogen (Nitrate/Nitrite), 1% (w/v), LabChem™
$155.32 - $222.89
Chemical Identifiers
| CAS | 63-74-1 |
|---|---|
| Molecular Formula | C6H8N2O2S |
| Molecular Weight (g/mol) | 172.20 |
| InChI Key | FDDDEECHVMSUSB-UHFFFAOYSA-N |
| Synonym | sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol |
| PubChem CID | 5333 |
| ChEBI | CHEBI:45373 |
| IUPAC Name | 4-aminobenzene-1-sulfonamide |
| SMILES | NC1=CC=C(C=C1)S(N)(=O)=O |
Chemical Identifiers
| 63-74-1 | |
| 172.20 | |
| sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol | |
| CHEBI:45373 | |
| NC1=CC=C(C=C1)S(N)(=O)=O |
| C6H8N2O2S | |
| FDDDEECHVMSUSB-UHFFFAOYSA-N | |
| 5333 | |
| 4-aminobenzene-1-sulfonamide |
Specifications
| Colorless | |
| 1,4.34,94.66 | |
| C6H8N2O2S | |
| sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol | |
| FDDDEECHVMSUSB-UHFFFAOYSA-N | |
| 4-aminobenzene-1-sulfonamide | |
| 5333 | |
| 172.2 | |
| Passes Test | |
| Hydrogen chloride; Carbon dioxide; Carbon monoxide; Nitrogen oxides; Sulfur compounds | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature