missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole USP , Medisca™
GREENER_CHOICE

Catalog No. 18048045
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048045 100 g
18048046 1 kg
18048108 500 g
18048133 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048045 Supplier Medisca Pharmaceutique Inc Supplier No. 026105

Sulfamethoxazole USP is a white to off-white, practically odorless, crystalline powder.

Chemical Identifiers

CAS 723-46-6

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White to Off-White
Quantity 100 g
Percent Purity 98.0 to 102.0%
Grade USP