missing translation for 'onlineSavingsMsg'
Learn More

Strontium ICP Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical

Catalog No. PSR1KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PSR1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSR1KN100 Supplier Ricca Chemical Company Supplier No. PSR1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10042-76-9 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sr
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.24
Equivalence 1 mL = 1 mg Sr
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Strontium ICP Standard
Show More Show Less