missing translation for 'onlineSavingsMsg'
Learn More

Strontium bromide hexahydrate, 99%

Numéro de catalogue. AAB2255322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAB2255322 100 g
AAB2255336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAB2255322 Fournisseur Thermo Scientific Chemicals Code fournisseur B2255322
Il en reste null

CAS: 7789-53-9 | Br2H12O6Sr | 355.518 g/mol

Strontium bromide hexahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Strontium bromide hexahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (2850 g/L at 20°C).

Notes
m.p. 88° (rapidly heated) , 180° -6H{2}O

Identifiants chimiques

CAS 7789-53-9
Molecular Formula Br2H12O6Sr
Molecular Weight (g/mol) 355.518
MDL Number MFCD00150361
InChI Key FLMJUJXBFKFYOZ-UHFFFAOYSA-L
Synonym unii-7i32n2ud6w, strontium dibromide hexahydrate, strontium bromide, reagent, strontium 2+ hexahydrate dibromide, strontium 2+ ion hexahydrate dibromide
PubChem CID 165644
IUPAC Name strontium;dibromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Sr+2]

Spécifications

Quantity 100 g
Linear Formula SrBr2·6H2O
Merck Index 14,8837
Solubility Information Soluble in water (2850g/L at 20°C).
Formula Weight 355.53 (247.44 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.386
Chemical Name or Material Strontium bromide hexahydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-969-5
RTECSNumber WK8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only