missing translation for 'onlineSavingsMsg'
Learn More

Strontium AA Standard, 1000 ppm Sr in 3% HCl, Ricca Chemical

Catalog No. ASR1KH500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ASR1KH500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ASR1KH500 Supplier Ricca Chemical Company Supplier No. ASR1KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1633-05-2 , 7647-01-0 , 7732-18-5
Molecular Formula CO3Sr
Molecular Weight (g/mol) 147.63
MDL Number MFCD00011250
InChI Key LEDMRZGFZIAGGB-UHFFFAOYSA-L
PubChem CID 15407
IUPAC Name strontium(2+) carbonate
SMILES [Sr++].[O-]C([O-])=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Sr
CAS Min % 94.0, 3.45, 0.16
CAS Max % 98.0, 3.59, 0.17
Equivalence 1 mL = 1 mg Sr
Physical Form Liquid
Color Colorless
Quantity 500 mL
Chemical Name or Material Strontium AA Standard