missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Sterile PET Media Bottles with HDPE Caps

Catalog No. S139079
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139079 125 mL
S139080 250 mL
S139081 500 mL
S139082 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S139079 Supplier Eisco Labs Supplier No. CH5221PK24

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sterile PET media bottles with secure HDPE caps, leakproof seals, and tamper-evident bands built for safe, reliable storage of lab reagents and buffers with lightweight durability and chemical resistance.

  • Sterile Media Bottles: PET bottles with secure HDPE caps, ideal for laboratory reagents and buffer storage.
  • Leakproof Design: Features semi-buttress threads and molded seal rings for a dependable, spill-free seal during storage and transport.
  • Tamper-evident Safety: Heat-shrink bands around each bottle ensure material security and protection from unauthorized access.
  • Durable and Lightweight: Made from robust PET with smooth, rounded edges for easy sample recovery and safe handling.
  • Wide Temperature Range: Suitable for storage between -70°C and +60°C, supporting diverse lab and biopharmaceutical applications.

Specifications

Capacity (Metric) 125 mL
Color Clear
Diameter (Metric) Inner Neck 28.5 mm
Material PET
No. per Pack 24 Pk.
Sterility Sterile
Neck Finish 52.3 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 107.4 mm
Autoclavable Not Autoclavable
Capacity (English) 4.22 oz.
Shape Cylindrical
Disposable Single-use
Product Type Media Bottle
Show More Show Less