missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Technology Station Charging and Storage Case

Catalog No. 17800653
Change view
Click to view available options
For Use With (Equipment):
CS3000 PAPR System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17800653 CS3000 PAPR System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17800653 Supplier CleanSpace™ Technology Supplier No. CS3014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Holds up to 8 units.

  • For use with HALO Power Systems
  • Holds up to 8 units

Specifications

Color Clear
Disposable Reusable
Product Type Station Charger/Case
For Use With (Equipment) CS3000 PAPR System
Model CS3014