Learn More
SciMatCo™ Stak-a-Cab™ Stackable Cabinet
Make your workspace work for you. SciMat™ Stak-a-CabSciMat™ allows you to design your own storage system and save valuable laboratory space.
$3,219.20
Spécifications
| Depth (Metric) Exterior | 46 cm |
|---|---|
| Color | Gray |
| Depth (English) Exterior | 18 in. |
| Dimensions (D x W x H) | 18 x 47 x 18 in. |
| Height (English) Exterior | 18 in. |
| Numéro de catalogue | Numéro du manufacturier. | Door Style | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Door Style | Prix | Quantity & Availability | |||||
|
19041859
|
SciMatCo™
SC2063 |
Self-close |
chaque for $3,219.20
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Customize storage with stackable poison, corrosives, flammables, and combination corrosives/flammables cabinets.
- Can be used singly on a benchtop or safely stacked up to three high with floor stand (sold separately)
- Feature liquid-tight trough floors, large hazard labels on both doors, and epoxy paint finish inside and out
- All SciMatCo flammables cabinets are UL listed
Stak-a-Cab Corrosives Cabinet
- No interior metal parts to corrode—has wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly
- Capacity: 22 x 2.5L (approximately 14 1/2 gal./55L)
- Will accept accessory nitric acid compartment (sold separately)
Stak-a-Cab Flammables Cabinet
- Includes two locks with two keys each and piano-style door hinges (all hardware is corrosion resistant)
- Capacity: 20 x 1 gal. (approximately 75.7L)
Stak-a-Cab Combination Cabinet
- Combination corrosives/flammables cabinet
- Corrosives compartment capacity: 10 x 2.5L (approximately 6 1/2 gal.)
- Flammables compartment capacity: 8 x 1 gal. (approximately 30L)
Stak-a-Cab Poison Cabinet
- Includes lock with two keys and piano-style door hinges (all hardware is corrosion resistant)
- Capacity: 16 x 1 gal. (approximately 60.5L)
Stak-a-Cab Floor Stands
- Accessory floor stands allow cabinets to be safely elevated from floor level: Fisher Cat: 17-988-486 (blue), 17-988-485 (yellow) and 17-988-487 (gray)
- Same epoxy paint finish as cabinets
- Height: 4.63 in. (11cm)
Order Info
Exterior dimensions: 18L x 47W x 18 in. H (46 x 119 x 46cm).
Compliance
Meet OSHA and NFPA requirements. Self-closing models meet UFC requirements.
Spécifications
| 46 cm | |
| 18 in. | |
| 18 in. | |
| Storage Cabinet | |
| Flammables | |
| 8 x 1 gal. flammable, 10 x 2.5 L Corrosive | |
| Epoxy |
| Gray | |
| 18 x 47 x 18 in. | |
| Wood (Plywood) | |
| 47 in. | |
| 45.7 cm | |
| 2 | |
| 119.4 cm |