missing translation for 'onlineSavingsMsg'
Learn More

SciMatCo™ Stak-a-Cab™ Stackable Cabinet

Catalog No. 19041859
Encompass
Change view
Click to view available options
Door Style:
Self-close
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
19041859 Self-close
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19041859 Supplier SciMatCo™ Supplier No. SC2063

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Make your workspace work for you. SciMat™ Stak-a-CabSciMat™ allows you to design your own storage system and save valuable laboratory space.

Customize storage with stackable poison, corrosives, flammables, and combination corrosives/flammables cabinets.

  • Can be used singly on a benchtop or safely stacked up to three high with floor stand (sold separately)
  • Feature liquid-tight trough floors, large hazard labels on both doors, and epoxy paint finish inside and out
  • All SciMatCo flammables cabinets are UL listed

Stak-a-Cab Corrosives Cabinet

  • No interior metal parts to corrode—has wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly
  • Capacity: 22 x 2.5L (approximately 14 1/2 gal./55L)
  • Will accept accessory nitric acid compartment (sold separately)

Stak-a-Cab Flammables Cabinet

  • Includes two locks with two keys each and piano-style door hinges (all hardware is corrosion resistant)
  • Capacity: 20 x 1 gal. (approximately 75.7L)

Stak-a-Cab Combination Cabinet

  • Combination corrosives/flammables cabinet
  • Corrosives compartment capacity: 10 x 2.5L (approximately 6 1/2 gal.)
  • Flammables compartment capacity: 8 x 1 gal. (approximately 30L)

Stak-a-Cab Poison Cabinet

  • Includes lock with two keys and piano-style door hinges (all hardware is corrosion resistant)
  • Capacity: 16 x 1 gal. (approximately 60.5L)

Stak-a-Cab Floor Stands

  • Accessory floor stands allow cabinets to be safely elevated from floor level: Fisher Cat: 17-988-486 (blue), 17-988-485 (yellow) and 17-988-487 (gray)
  • Same epoxy paint finish as cabinets
  • Height: 4.63 in. (11cm)

Order Info

Exterior dimensions: 18L x 47W x 18 in. H (46 x 119 x 46cm).

Compliance

Meet OSHA and NFPA requirements. Self-closing models meet UFC requirements.

Specifications

Product Type Storage Cabinet
Load Bearing Capacity 8 x 1 gal. flammable, 10 x 2.5 L Corrosive
Door Style Self-close
Dimensions (D x W x H) 18 x 47 x 18 in.
Height (Metric) Exterior 45.7 cm
Width (Metric) Exterior 119.4 cm
No. of Doors 2
Depth (Metric) Exterior 46 cm
Height (English) Exterior 18 in.
Certifications/Compliance NFPA, OSHA, UFC
Material Wood (Plywood)
Includes Internal Metal lock on the flammable side and a Plastic hasp and padlock on the corrosive side
Depth (English) Exterior 18 in.
Color Gray
Surface Coating Epoxy
For Use With (Application) Flammables
Width (English) Exterior 47 in.
Show More Show Less