missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3192 Wrist Lanyard Tethering Kit - 3lbs/1.4kg

Catalog No. 19910460
Change view
Click to view available options
Load Bearing Capacity (Metric):
1.4 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (Metric)
19910460 1.4 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19910460 Supplier Ergodyne™ Supplier No. 19662

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tool Tethering Kit allows users to tether tools, making it easy to use tools on the job while preventing dropped objects when working at-heights.

  • Allows for tethering up to four 3 lb. / 1.4 kg tools
  • For: pliers, claw hammers, sledge hammers, cat's paws, utility knives, jab/drywall saws, caulking knives, nips and trowels
  • Straightforward installation
  • Lack of tool lanyards can create a hazardous environment while working at heights
  • Measure and cut the tape to the specified length listed in instructions. Lay the tool tail on the tool being tethered and wrap the tape once around itself beginning directly below the barrel lock. Continue wrapping and stretching the tape firmly. Firmly push down on the end and attach the Squids™ Pull-On Wrist Lanyard to complete the tethering system

Specifications

Certifications/Compliance ANSI/ISEA 121-2018
Load Bearing Capacity (Metric) 1.4 kg
Includes Squids™ 3116 Pull-On Wrist Lanyard with Buckle, a 12ft / 3.7m roll of Squids™ 3755 Self-Adhering Tape Trap, and three Squids™ 3103 Detachable Elastic Tool Tails
Load Bearing Capacity (English) 3 lb.