missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Squids™ 3100F(x) Tool Lanyard Single Carabiner

Catalog No. 19910461
Change view
Click to view available options
Load Bearing Capacity (English):
10 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity (English)
19910461 10 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19910461 Supplier Ergodyne™ Supplier No. 19802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Squids™ 3100F(x) Tool Lanyard Single Carabiner is designed to prevent falling objects and reduce shock load.

  • Approved to the ANSI/ISEA 121-2018 standard
  • Shock absorbing properties reduce the dynamic force on the body or other anchor point
  • Durable 1680 denier polyester elastic blended webbing extends
  • Low profile lightweight design reduces snag hazards and added weight
  • Anodized aluminum alloy carabiner with captive eye
  • Treaded loop end that grips the tool when attached
  • High strength 3-ply nylon stitching secures connecting ends
  • Single length construction eliminates weak points seen with multi-component lanyards
  • Certified to a 2:1 safety factor

Specifications

Color Orange, Gray
Load Bearing Capacity (Metric) 4.5 kg
Load Bearing Capacity (English) 10 lb.
Length (Metric) 96 to 122 cm
Certifications/Compliance ANSI/ISEA 121-2018 standard
Length (English) 38 to 48 in.