missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals SPINeasy™ 96-Well DNA Pro Kit for Soil
Designed in 96-well spin plate format, allowing for high throughput and rapid sample processing.
$1,412.60 - $4,340.88
Specifications
| Format | Spin Column |
|---|---|
| Isolation Technology | Silica Column |
| Scale | Mini |
| Product Type | DNA Pro Kit for Soil |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|
MP116546096
|
MP Biomedicals
116546096 |
96 Preps |
Pack for $1,412.60
|
|
|||||
|
MP116546496
|
MP Biomedicals
116546496 |
4 x 96 Preps |
Pack for $4,340.88
|
|
|||||
Description
- Enables efficient lysis of microbial population present in complex soil samples, including bacteria, fungi, viruses, and protists ensuring comprehensive genomic DNA extraction.
- Compatible with various challenging soil types, including those with low biomass or high contamination levels.
- Streamlined workflow reduces processing time, enabling faster and convenient isolation of pure microbial genomic DNA in 90 minutes.
Specifications
| Spin Column | |
| Silica Column | |
| Mini | |
| DNA Pro Kit for Soil |