Learn More
MP Biomedicals SPINeasy™ 96-Well DNA Pro Kit for Soil
Designed in 96-well spin plate format, allowing for high throughput and rapid sample processing.
Supplier: MP Biomedicals 116546496
Description
- Enables efficient lysis of microbial population present in complex soil samples, including bacteria, fungi, viruses, and protists ensuring comprehensive genomic DNA extraction.
- Compatible with various challenging soil types, including those with low biomass or high contamination levels.
- Streamlined workflow reduces processing time, enabling faster and convenient isolation of pure microbial genomic DNA in 90 minutes.
Chemical Identifiers
| Spin Column | |
| Silica Column | |
| Mini | |
| DNA Pro Kit for Soil |
Specifications
| Spin Column | |
| Mini | |
| DNA Pro Kit for Soil |
| Silica Column | |
| 4 x 96 Preps |