Learn More
MP Biomedicals SPINeasy™ 96-Well DNA Pro Kit for Soil
Designed in 96-well spin plate format, allowing for high throughput and rapid sample processing.
Supplier:  MP Biomedicals 116546096
 View more versions of this product
Catalog No. MP116546096
Only null left
Description
Description
  • Enables efficient lysis of microbial population present in complex soil samples, including bacteria, fungi, viruses, and protists ensuring comprehensive genomic DNA extraction.
  • Compatible with various challenging soil types, including those with low biomass or high contamination levels.
  • Streamlined workflow reduces processing time, enabling faster and convenient isolation of pure microbial genomic DNA in 90 minutes.
Specifications
Chemical Identifiers
Spin Column
Silica Column
Mini
DNA Pro Kit for Soil
Specifications
Spin Column
Mini
DNA Pro Kit for Soil
Silica Column
96 Preps
Product Suggestions
Product Suggestions