missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™
Description
Comprend
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Al | 200μg/mL |
| As | 200μg/mL |
| Ba | 200μg/mL |
| Se | 200μg/mL |
| Tl | 200μg/mL |
| Fe | 100μg/mL |
| Co | 50μg/mL |
| Mn | 50μg/mL |
| Ni | 50μg/mL |
| Pb | 50μg/mL |
| Sb | 50μg/mL |
| V | 50μg/mL |
| Zn | 50μg/mL |
| Cu | 25μg/mL |
| Cr | 20μg/mL |
| Ag | 5μg/mL |
| Be | 5μg/mL |
| Cd | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
| Chemical Name or Material | ICP Spike Sample 1 |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | GC-MS |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | Components in 5% HNO3/tr Tartaric Acid/tr HF: Al •200μg/mL Sb •50μg/mL As •200μg/mL Ba •200μg/mL Be •5μg/mL Cd •5μg/mL Cr •20μg/mL Co •50μg/mL Cu •25μg/mL Fe •100μg/mL Pb •50μg/mL Mn •50μg/mL Ni •50μg/mL Se •200μg/mL Ag •5μg/mL Tl •200μg/mL V •50μg/mL Zn •50μg/mL |
Product intended for laboratory use only