missing translation for 'onlineSavingsMsg'
Learn More

3M™ Speedglas™ Inside Protection Plate

Catalog No. 296368088
Change view
Click to view available options
Visor Type:
Variable 5, 8-13
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Visor Type
296368088 Variable 5, 8-13
296368086 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296368088 Supplier 3M™ Supplier No. 7000127165

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to protect the inside surface of the filter from spark and spatter

  • Protects interior of filter from spark and spatter
  • Helps prevent damage to the glass

Specifications

Color Clear
For Use With (Application) Assembly, Facility Maintenance, Grinding, Machine Operations, Sanding, Welding
Product Type Replacement Parts and Accessories
Quantity 25/Cs.
Visor Type Variable 5, 8-13