Learn More
Epredia™ Specialized sandwich supporting material
Ensure accuracy and efficiency with Epredia™ Specialized Clamps and Accessories.
Supplier: Epredia™ 715030
Description
- For clamping thin materials
- Spring-loaded jaws secure specimen
- Compensation slot ensures correct specimen clamping
- Orienting on the Z-axis
- Foil clamp is fitted directly on the specimen cylinder
- Sandwich supporting material is for foil clamp to section thin foil specimens; 50 squares of hard material and two strips
Identifiants chimiques
| Epredia Clamps |
Spécifications
| Foil Clamp | |
| Epredia Clamps |