Learn More
Epredia™ Specialized sandwich supporting material
Catalog No. p-4529868
Change view
Click to view available options
Product Type:
Foil Clamp
Catalog No. Product Type
361013427 Foil Clamp
1 options
Catalog No. 361013427 Supplier Epredia™ Supplier No. 715030
Only null left
Ensure accuracy and efficiency with Epredia™ Specialized Clamps and Accessories.
  • For clamping thin materials
  • Spring-loaded jaws secure specimen
  • Compensation slot ensures correct specimen clamping
  • Orienting on the Z-axis
  • Foil clamp is fitted directly on the specimen cylinder
  • Sandwich supporting material is for foil clamp to section thin foil specimens; 50 squares of hard material and two strips
Specifications
Product Type Foil Clamp
For Use With (Equipment) Epredia Clamps