Learn More
Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Constructed of Durastar™ co-polyester. Standard desiccator cabinets block 99% of UV light and are resistant to staining, crazing, and chemical attack.
Supplier:  Bel Art™ F420710000
 Afficher plus de versions de ce produit
Numéro de catalogue. 0864750
Get More Bel-Art Lab Products with Purchase
Afficher toutes les promotions
Il en reste null
Description
Description
  • Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Stack up to 3 units high to save space
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Spécifications
Identifiants chimiques
Not Autoclavable
Copolyester
Left-hinged and gasketed
1.0
Chemicals, stains and fine cracks
0.75 cu. ft.
16.3 in.
13.4 in.
34.1 cm
4.5 in.
16.3 in.
Vertical
Desiccator
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
1
Fixed Perforated
16.3 x 13.4 in. (41.4 x 34.1 cm)
41.4 cm
8.9 in.
8.4 in.
21.3 cm
41.4 cm
Spécifications
Not Autoclavable
Vertical
Desiccator
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
1.0
Chemicals, stains and fine cracks
0.75 cu. ft.
16.3 in.
13.4 in.
34.1 cm
8.4 in.
21.3 cm
41.4 cm
Clear
Copolyester
Left-hinged and gasketed
Carrying handles, one latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer
1
Fixed Perforated
16.3 x 13.4 in. (41.4 x 34.1 cm)
41.4 cm
8.9 in.
22.5 cm
4.5 in.
16.3 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS