missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Constructed of Durastar™ co-polyester. Standard desiccator cabinets block 99% of UV light and are resistant to staining, crazing, and chemical attack.
$1,161.21 - $1,283.39
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Cabinet Style | Vertical |
| Material | Copolyester |
| Product Type | Desiccator |
| Door Style | Left-hinged and gasketed |
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
08647850
|
Bel Art™
F420710008 |
Amber |
Each for $1,161.21
|
|
|||||
|
0864750
|
Bel Art™
F420710000 |
Clear |
Each for $1,283.39
|
|
|||||
Description
- Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Stack up to 3 units high to save space
- Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications
| Not Autoclavable | |
| Copolyester | |
| Left-hinged and gasketed | |
| 1.0 | |
| Chemicals, stains and fine cracks | |
| 0.75 cu. ft. | |
| 16.3 in. | |
| 13.4 in. | |
| 34.1 cm | |
| 4.5 in. | |
| 16.3 in. |
| Vertical | |
| Desiccator | |
| Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 | |
| 1 | |
| Fixed Perforated | |
| 16.3 x 13.4 in. (41.4 x 34.1 cm) | |
| 41.4 cm | |
| 8.9 in. | |
| 8.4 in. | |
| 21.3 cm | |
| 41.4 cm |