missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol Solution, USP, 70%, Spectrum™ Chemical

SO220, 50-70-4, C6H14O6

Supplier:  Spectrum Chemical Mfg Cor SO2204LTGLCS4

Quantity 4 x 4 L
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18606953

Only null left

Description

Description

Spectrum™ Chemical Sorbitol Solution, 70 Percent, USP is a hyperosmotic laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

50-70-4 , 7732-18-5
182.17
hexane-1,2,3,4,5,6-hexol
C6H14O6
FBPFZTCFMRRESA-UHFFFAOYNA-N
OCC(O)C(O)C(O)C(O)CO

Specifications

50-70-4 , 7732-18-5
0.001
C6H14O6
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
0.7, 0.3
Amber Glass Bottle
4 x 4 L
OCC(O)C(O)C(O)C(O)CO
182.17
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions