missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol Solution, USP, 70%, Spectrum™ Chemical
SO220, 50-70-4, C6H14O6
$1,088.24 - $14,696.70
Chemical Identifiers
| CAS | 50-70-4 , 7732-18-5 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606951
|
N/A | 6 x 500 mL | Amber Glass Bottle |
Case of 6 Each for $1,088.24
|
|
||||
|
18606953
|
N/A | 4 x 4 L | Amber Glass Bottle |
Case of 4 Each for $3,228.70
|
|
||||
|
18606954
|
N/A | 20 L | Poly Drum |
Each for $2,942.06
|
|
||||
|
18606955
|
N/A | 200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||