Learn More
3M™ Solus™ 1000 Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Catalog No. 19169047
Change view
Click to view available options
Product Type:
Safety Glasses
Safety Glasses Kit
Frame Color:
Blue, Black
Lens Tint:
Amber
Clear
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Frame Color Lens Tint
19169047 Safety Glasses Kit Blue, Black Clear
19169044 Safety Glasses Blue, Black Clear
19169046 Safety Glasses Blue, Black Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19169047 Supplier 3M™ Supplier No. 7100079186
Only null left
3M™ Solus™ Safety Glasses, feature a clear, strong polycarbonate lens in a slim, stylish, green and black-accented colour frame. Our safety glasses have 3M™ Scotchgard™ Anti-Fog / Anti-Scratch Coating, and a soft temple and nose bridge.
  • Stylish and modern look slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Lightweight with soft cushioning on nose bridge
  • Inner padding on temples for increased comfort and stability when worn
  • Meets K&N marking requirements of EN 166
  • Frame marking: 3M EN166 FT CE
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Certifications/Compliance ANSI Z87.1+ 2015, CSA Z94.3
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Glasses Kit
For Use With (Application) Construction, Food and Beverage Manufacturing, General Manufacturing, Metal Fabrication, Military Maintenance, Repair and Operation (MRO), Oil & Gas, Primary Metals, Transportation
Frame Color Blue, Black
Includes Removable foam gasket and elastic strap
Model S1101SGAF-KT
Temple Style Removable
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Half Frame
Show More Show Less