missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Solid Top Screw Cap with PTFE Liner

Numéro de catalogue. p-7213898
Change view
Click to view available options
For Use With (Equipment):
15 mL Vial
20 or 40 mL Vial
22 mL Vial
7 mL Vial
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. For Use With (Equipment)
111020923 7 mL Vial
111010572 20 or 40 mL Vial
111011290 15 mL Vial
111011291 22 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 111020923 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 27152
Il en reste null

Known for providing the widest range of chemical compatibility and for being the most temperature tolerant of plastic closures

  • The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate.
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene).
  • Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents. 

Spécifications

Material (Liner) F217/PTFE
Color Green
Material Melamine Resin
No. per Pack 100
Closure Size 15-425
For Use With (Equipment) 7 mL Vial
Product Type Solid Cap