missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Solid Top Screw Cap with Aluminum Liner

Numéro de catalogue. 111012040
Change view
Click to view available options
For Use With (Equipment):
15 mL Vial
2 mL Standard Opening Vial
4 mL Vial
7 mL Vial
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. For Use With (Equipment)
111012040 7 mL Vial
111012036 2 mL Standard Opening Vial
111012038 4 mL Vial
111012041 15 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 111012040 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 27153
Il en reste null

Available for use with sealing septa and solid caps with aluminum liners

  • Caps are offered in solid phenolic caps that can withstand a maximum temperature of 135°C.
  • Screw caps are available as solid caps with aluminum liner.
  • Caps are made from phenolic resin.
  • Screw caps are generally available as solid caps for use with sealing septa.

Spécifications

Material (Liner) Aluminum
Color Black
No. per Pack 100
Closure Size 15-425
For Use With (Equipment) 7 mL Vial
Product Type Phenolic Solid Cap