missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solid Rubber Stoppers

Available in a variety of sizes ranging from 000 through 16

$37.75 - $133.18

Specifications

Color Black
Material Natural Rubber
Product Type Solid Stopper
For Use With (Application) General purpose use
View More Specs

Products 22
Catalog Number Mfr. No. Size Stopper Number Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size Stopper Number Length (Metric) Price Quantity & Availability  
14130S
View Documents Promotion Details
N/A 11.5 11.5 2.5 cm
Pack of 5 Each for $107.00
Case of 5 Pack for $535.00
Only null left
 
14130E
View Documents Promotion Details
N/A 3 3 2.5 cm
Pack of 39 Each for $79.39
Case of 5 Pack for $396.95
Only null left
 
14130R
View Documents Promotion Details
N/A 11 11 2.5 cm
Pack of 6 Each for $104.75
Case of 5 Pack for $523.75
Only null left
 
14130B
View Documents Promotion Details
N/A 0 0 2.5 cm
Pack for $74.75
Case of 5 Pack for $373.75
Only null left
 
14130D
View Documents Promotion Details
N/A 2 2 2.5 cm
Pack of 52 Each for $82.21
Case of 5 Pack for $411.05
Only null left
 
14130N
View Documents Promotion Details
N/A 9 9 2.5 cm
Pack of 10 Each for $101.68
Case of 5 Pack for $508.40
Only null left
 
14130H
View Documents Promotion Details
Fisherbrand™
JUI5.5SOL
5.5 5.5 2.5 cm
Pack of 26 Each for $42.39
Case of 5 Pack for $211.95
Only null left
 
141351A
View Documents Promotion Details
N/A 000 0 2.1 cm
Pack of 181 Each for $133.18
Case of 5 Pack for $665.90
Only null left
 
14130G
View Documents Promotion Details
N/A 5 5 2.5 cm
Pack of 28 Each for $87.43
Case of 5 Pack for $437.15
Only null left
 
14130V
View Documents Promotion Details
N/A 14 14 3.9 cm
Each for $59.39
Case of 8 Each for $475.12
Only null left
 
14130W
View Documents Promotion Details
N/A 15 15 3.9 cm
Each for $72.57
Case of 8 Each for $580.56
Only null left
 
14130P
View Documents Promotion Details
N/A 10 10 2.5 cm
Pack of 8 Each for $91.46
Case of 5 Pack for $457.30
Only null left
 
14130F
View Documents Promotion Details
N/A 4 4 2.5 cm
Pack of 33 Each for $83.68
Case of 5 Pack for $418.40
Only null left
 
14130UU
View Documents Promotion Details
N/A 13.5 13.5 3.5 cm
Each for $37.75
Case of 13 Each for $490.75
Only null left
 
14130M
View Documents Promotion Details
N/A 8 8 2.5 cm
Pack of 12 Each for $82.89
Case of 5 Pack for $414.45
Only null left
 
14130A
View Documents Promotion Details
N/A 00 0 2.5 cm
Pack for $77.29
Case of 5 Pack for $386.45
Only null left
 
14130U
View Documents Promotion Details
N/A 13 13 2.5 cm
Pack of 4 Each for $94.79
Case of 5 Pack for $473.95
Only null left
 
14130Q
View Documents Promotion Details
N/A 10.5 10.5 2.5 cm
Pack of 7 Each for $74.39
Case of 5 Pack for $371.95
Only null left
 
14130C
missing translation for 'viewDocuments' Promotion Details
N/A 1 1 2.5 cm
Pack for $78.54
Case of 5 Pack for $392.70
Only null left
 
22033987
missing translation for 'viewDocuments' Promotion Details
N/A 9.5 9.5 2.5 cm
Pack of 9 Each for $66.39
Case of 5 Pack for $331.95
Only null left
 
14130L
missing translation for 'viewDocuments' Promotion Details
N/A 7 7 2.5 cm
Pack of 15 Each for $81.75
Case of 5 Pack for $408.75
Only null left
 
14130T
missing translation for 'viewDocuments' Promotion Details
N/A 12 12 2.5 cm
Pack of 5 Each for $99.64
Case of 5 Pack for $498.20
Only null left
 
Description

Description

Available in a variety of sizes ranging from 000 through 16

  • Solid rubber construction

The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).

Specifications

Specifications

Black
Natural Rubber
Solid Stopper
General purpose use
Videos
SDS
Documents

Documents